missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 53-001 Nitrile Chemical Resistant Gloves
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Click to view available options
Size:
7
8
9
10
11
Description
- Innovative multi-layer polymer design - nitrile/neoprene/nitrile layers
- Provides chemical protection against a wide range of chemicals from acids and bases to hydrocarbons and organic solvent
- Tested according to EN ISO 374:2016, permeation time > 30 minutes against 13 chemicals out of 18 listed
- Features the latest MICROCHEM™ chemical barrier technology providing superior protection for use in hazardous environments
- ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
- Inner soft nylon liner for comfort and increased mechanical protection
- Oeko-Tex™ Standard 100 Approved
Specifications
ANSI Abrasion Rating | ANSI 3 |
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, EN 1149, EN407 |
Chlorinated | No |
Color | Gray, Black |
Latex Free | Yes |
Material | Nitrile, Neoprene |
Size | 9 |
Thickness (English) | 15 mil |
Thickness (Metric) | 0.38 mm |
EN388 Standard | EN 388:3121A |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'