Learn More
Ansell AlphaTec™ 37-210 Reusable Nitrile Gloves with UltraGrip™ Technology
UltraGrip™ Technology delivers an innovative raised diamond grip on the glove's surface for enhanced grip capabilities in wet and oily conditions.
£19.15
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Color Blue
Disposable Reusable
Hand Specific
View More Specs
Products 6 Description
Description
  • Reusable nitrile gloves designed specifically for food processing and handling
  • Raised diamond-pattern enhances grip in wet and oily conditions
  • Gloves maintain flexibility and dexterity, even at low temperatures
  • Use alone or as outer glove over thermal liners or cut-resistant gloves
  • Latex-free formulation presents no risk of Type 1 allergies
Specifications
Specifications
ANSI 4
Blue
Specific
Nitrile
3001X
JKT
EN 420:2003 + A1:2009, REACH Compliant
Reusable
Yes
8 mil
Type B
37-210
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell AlphaTec™ 37-210 Reusable Nitrile Gloves with UltraGrip™ Technology