Learn More
Astell Scientific™ Air Ballast
Reduce the risk of bottled fluids boiling over. Astell Scientific™ Air Ballast requires compressed air.
Brand: Astell Scientific™ AVC004E
Additional Details : Weight : 0.01000kg
Description
- Air ballast reduces the risk of bottled fluids boiling over when an efficient cooling system (e.g., water jacket cooling) causes a sudden change in pressure within the autoclave chamber.
- Allows the cooling stage to be implemented earlier, which reduces cooling time.
- Requires compressed air system.
Specifications
Requires compressed air system | |
Air Ballast |