Learn More
Beckman Coulter™ Agencourt AMPure XP
Trusted and recommended reagent for size selection and cleanup steps within a variety of NGS workflows. Beckman Coulter™ Agencourt AMPure XP removes unwanted contaminants from DNA in a variety of applications including PCR, NGS, cloning, and microarrays.
Brand:  Beckman Coulter™ A63882
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15522534
  • £8046.00 / 450mL
Estimated Shipment: 25-07-2024
to see stock.
Description
Description
  • SPRI paramagnetic bead-based chemistry
  • Removes contaminants (dNTPs, salts, primers, primer dimers)
  • High recovery of amplicons (>100 bp) ensures that critical data is not lost
  • Trusted and recommended reagent for a variety of NGS workflows
  • Compatible with manual and automated processing
  • Predictable and consistent size selection
  • Manual cleanups using AMPure XP require less than 30 minutes
  • AMPure XP is automated on all Biomek platforms for higher throughput
Specifications
Specifications
AMPure XP, Speed Beads (Mag Carboxy)
Liquid
8.0 to 8.4
DNA
2-8°C
Automated 96- or 384-well formats, e.g. Biomek platforms
Standard PCR, Real Time PCR (qPCR), Next-Generation Sequencing (NGS)
450mL