missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose, Molecular biology reagent grade, MP Biomedicals™
Click to view available options
Quantity:
100 g
500 g
Specifications
Physical Form | Powder |
Color | White |
Quantity | 100 g |
Grade | Molecular Biology |
Chemical Name or Material | Agarose |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'