Learn More
B Braun™ Aesculap™ Favorita™ Blades
High-quality carbon steel blades with recognized quality for professional use.
POA
Specifications
For Use With (Application) | Cutting/Shaving Hair |
---|---|
Type | Clipper Blades |
For Use With (Equipment) | Aesculap™ Favorita™ clippers |
Material | Carbon Steel |
Description
- High-quality carbon steel blades offer exceptionally long cutting edge retention
- Quality workmanship – all parts are milled or profile-ground
- Adjustable plate pressure
- High-end surface finish against corrosion
- Multiple sharpening service
Specifications
Cutting/Shaving Hair | |
Clipper Blades | |
Aesculap™ Favorita™ clippers | |
Carbon Steel |