missing translation for 'onlineSavingsMsg'
Learn More

Adrenomedullin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10778-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887383

  • £339.00 / 150µL
Estimated Shipment: 05-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintece of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
Specifications

Specifications

Adrenomedullin
Polyclonal
Unconjugated
ADM
ADM, adrenomedullin, AM, PAMP, ProAM N terminal 20 peptide, ProAM N20
Rabbit
Antigen Affinity Chromatography
RUO
133
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P35318
ADM
Adrenomedullin Fusion Protein Ag1197
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers