missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine, 99%

Product Code. 11314619
Click to view available options
Quantity:
25 g
50 g
250 g
500 g

Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

Solubility
Soluble in hot water, methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Melting Point >360°C (decomposition)
Assay Percent Range 99%
Odor Odorless
Quantity 250 g
UN Number UN2811
Beilstein 5777
Merck Index 14,152
Solubility Information Soluble in hot water,methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.
Formula Weight 135.13
Percent Purity 99%
Chemical Name or Material Adenine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'