Learn More
HUBER Adapters
Accessories
Brand:  HUBER 6447
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10495912
  • £143.27 / Each
Estimated Shipment: 18-07-2024
to see stock.
Specifications
M30 x 1.5 male thread to G3/4 male thread
1 Ea.
0.130 kg
Adapter