missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Acura™ Electro 926 Electronic Single-Channel Micropipette Initial Kit

Ergonomic electronic single-channel micropipette initial package. Acura™ Electro 926 Electronic Single-Channel Micropipette Initial Kit includes pipette, charging stand, fast charger, AC adapter and cord, additional battery pack, and pipette tip samples.

Brand:  Socorex™ 926.0050E

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11899213

  • £795.00 / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket

Description

Description

  • Exceptional, user-friendly ergonomics.
  • Lightweight.
  • Easy, intuitive, self-teaching program.
  • Large display, reversible right/left reading.
  • Microprocessor-controlled motor.
  • Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
  • Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
  • Includes pipetting cycle counter.
  • Optional volumetric modules.
Specifications

Specifications

<±1.0% to <±0.5%
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS)
1
Starter Kit, Micro
Right/Left LCD
0.25μL
1 Ea.
24 Months
4.8 V NiMH
926
Acura
2.5 to 50 μL
Pipette charging stand, fast charger with AC adapter and cord, additional battery pack, QC certificate, Qualitips™ pipette tip samples and operating instructions.
<0.7% to <0.25%
<1.5 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers