Learn More
Socorex™ Acura™ Electro 926 Electronic Single-Channel Micropipette Initial Kit
Ergonomic electronic single-channel micropipette initial package. Acura™ Electro 926 Electronic Single-Channel Micropipette Initial Kit includes pipette, charging stand, fast charger, AC adapter and cord, additional battery pack, and pipette tip samples.
Brand: Socorex™ 926.0002E
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
- Includes pipetting cycle counter.
- Optional volumetric modules.
Specifications
<±2.5% to <±0.9% | |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
1 | |
Starter Kit, Micro | |
Right/Left LCD | |
0.01 μL | |
1 Ea. | |
24 Months |
4.8 V NiMH | |
926 | |
Acura | |
0.1 to 2.0 μL | |
Pipette charging stand, fast charger with AC adapter and cord, additional battery pack, QC certificate, Qualitips™ pipette tip samples and operating instructions. | |
<2.5% to <0.8% | |
<1.5 hr. |