missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Versapor™ Membrane

For use in cell culture and other sensitive biological research.

Brand:  Cytiva (formerly Pall Lab) 4189

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 17154271

  • £193.00 / Pack of 75
Estimated Shipment: 24-10-2024
to see stock.
Add to basket

Description

Description

  • Meets aqueous sample filtration needs
  • Designed for laboratory applications only
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

Acrylic Copolymer (Hydrophilic)
Non-sterile
<70 μL
300/Cs.
55°C
0.8 μm
2.8 cm2
Modified Acrylic
75/Pk.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers