Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Versapor™ Membrane
For use in cell culture and other sensitive biological research.
Brand: Cytiva (formerly Pall Lab) 4473
Additional Details : Weight : 1.32050kg
Description
- Meets aqueous sample filtration needs
- Designed for laboratory applications only
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Acrylic Copolymer (Hydrophilic) | |
Non-sterile | |
4 mm |
0.45 μm | |
1000/Pk. |