missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with PTFE Membrane and Male Slip Luer Outlet

Universal membrane syringe filters for all of your analytical filtration requirements.

Brand:  Cytiva (formerly Pall Lab) 4422

Additional Details : Weight : 0.63650kg

 View more versions of this product

Product Code. 17184301

  • £251.81 / Pack of 100
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

PTFE
Non-sterile
<30 μL
300/Cs.
Male Slip Luer Outlet
13 mm
0.45 μm
1 cm2
Polypropylene
100/Pk.
100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers