missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
107.84 GBP valid until 2024-05-31
Use promo code "21688" to get your promotional price.
107.84 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.
107.84 GBP valid until 2024-12-31
Use promo code "21905" to get your promotional price.
Use promo code "21688" to get your promotional price.
107.84 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.
107.84 GBP valid until 2024-12-31
Use promo code "21905" to get your promotional price.
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Poly Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -94°C |
Appearance | Colorless liquid |
Density | 0.791g/mL at 25°C |
Boiling Point | 56°C (760 mmHg) |
Quantity | 6 x 1 L |
Explosion Limit | 0.132 |
Flash Point | −17°C (1.4°F) |
Packaging | Poly Bottle |
Refractive Index | n20/D 1.359 |
Residue after Evaporation | 0 to 0.001% |
Show More |
Safety and Handling
Product Identifier
- Acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'