missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™

Product Code. 15673920 Shop All Honeywell Fluka Products
200 L, Poly Drum
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
60 kg
6 x 1 L
200 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic Pail
Poly Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.2°C (literature)
Boiling Point 118°C
CAS Max % 100.0000%
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05
Percent Purity >99.8%
Grade ACS Reagent/Reagent ISO/Reagent Ph. Eur.
Density 1.049 g/mL at 25 °C(lit.)
Quantity 200 L
Explosion Limit 16% at 92°F, 4% at 59°F
Flash Point 40°C
Ignition Point 800°F
Packaging Poly Drum
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.