missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, 99%, Thermo Scientific Chemicals

Catalog Number p-7020582
Click to view available options
Quantity:
250 g
1000 g
5000 g
60-35-5
C2H5NO
59.068
MFCD00008023
DLFVBJFMPXGRIB-UHFFFAOYSA-N
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
178
CHEBI:49028
acetamide
CC(=O)N
60-35-5
C2H5NO
59.068
MFCD00008023
DLFVBJFMPXGRIB-UHFFFAOYSA-N
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
178
CHEBI:49028
acetamide
CC(=O)N

Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamide is used as an industrial solvent, plasticizer, wetting and penetrating agent. It is also used for the transamidation of carboxamides in 1,4-dioxane. It finds application in lacquers and soldering flux. Further, it acts as a precursor to thioacetamide.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 76°C to 81°C
Density 1.159
Boiling Point 220°C to 222°C
Quantity 250 g
Beilstein 1071207
Sensitivity Hygroscopic
Merck Index 14,43
Solubility Information Soluble in water.
Formula Weight 59.07
Percent Purity 99%
Chemical Name or Material Acetamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'