missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaldehyde dimethyl acetal, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.
Solubility
Soluble in water (1000 mg/ml at 25°C), alcohols, chloroform, ether, and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.
Solubility
Soluble in water (1000 mg/ml at 25°C), alcohols, chloroform, ether, and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | -113°C |
Density | 0.852 |
Boiling Point | 63°C to 65°C |
Flash Point | −17°C (1°F) |
Odor | Sweet |
Refractive Index | 1.367 |
Quantity | 500 g |
UN Number | UN2377 |
Beilstein | 1697039 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'