Learn More
Ansell AccuTech™ 91-250 Series Natural Rubber Latex Gloves Product Code.: 11791593
Ansell AccuTech™ 91-250 Series Natural Rubber Latex Gloves
Product Code. 11791593 Shop All Ansell Products
Click to view available options
Size:
6.5
7.5
8
8.5
9
Product Code. 11791593
Brand: Ansell 91250070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide optimal solutions for controlled environments. Low allergenic content minimizes the risk of latex associated allergies for the wearer.
  • Manufactured using a specific formulation of latex to offer users an increased level of flexibility and elasticity
  • Inner silicone coating facilitates easy donning and double-gloving
  • Thicker cuffs fit nicely over garments
TRUSTED_SUSTAINABILITY
Specifications
Cleanroom Class Class 100 (ISO 5)
Disposable Single-use
Latex Free No
Product Type Cleanroom Gloves
Quantity 1 Pack
Product Content Correction
Product Title
Ansell AccuTech™ 91-250 Series Natural Rubber Latex Gloves >