missing translation for 'onlineSavingsMsg'
Learn More
Ansell AccuTech™ 91-250 Series Natural Rubber Latex Gloves Product Code.: 11711603
Add to basket

Ansell AccuTech™ 91-250 Series Natural Rubber Latex Gloves

Product Code. 11711603 Shop All Ansell Products
8
Click to view available options
Size:
6.5
7.5
8
8.5
9

Product Code. 11711603

Brand: Ansell 91250080

Edge
Add to basket
Add to basket

Provide optimal solutions for controlled environments. Low allergenic content minimizes the risk of latex associated allergies for the wearer.

  • Manufactured using a specific formulation of latex to offer users an increased level of flexibility and elasticity
  • Inner silicone coating facilitates easy donning and double-gloving
  • Thicker cuffs fit nicely over garments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001, ISO 13485, CE Mark & ISO 11137
Cleanroom Class Class 100 (ISO 5)
Color Natural
Disposable Single-use
Latex Free No
Material Latex
Palm Thickness (Metric) 0.18 mm
Powdered Powder Free
Product Type Cleanroom Gloves
Size 8
Sterility Sterile
For Use With (Application) Biotechnologies, analytics testing, blistering operations, pharmaceuticals
Glove Cuff Color Natural
Glove Cuff Style Straight
Inspected for Medical Use Yes
Quantity 200 Pairs
Series 91-250
Length (Metric) 28.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell AccuTech™ 91-250 Series Natural Rubber Latex Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.