Learn More
Thermo Scientific™ Accessories for the LP Vortex Mixer
Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.
Marca: Thermo Scientific™ 88880122
Code : NEW
Detalles adicionales : Weight : 0.11000kg
Descripción
Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.
- Warranty: 2 years
Especificaciones
Tube Holder | |
15 mm | |
Tube Holder with Insert Retainer (dia. 15mm) | |
Vortex Mixer |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.