Learn More
Macherey-Nagel™ Accessories for Stainless Steel HPLC Columns
Macherey-Nagel™ Accessories for Stainless Steel HPLC Columns include: plastic column end caps, capillary unions (100mm x 1/16 in. x 0.25mm), 1/16 in. nuts for connecting capillaries, and 1/16 in. ferrules.
£30.90 - £95.55
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10082691
|
Macherey-Nagel™
718582 |
Locking Screw |
£30.90
Pack of 4 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
10543412
|
Macherey-Nagel™
718583 |
Terminal Screw |
£34.95
Pack of 5 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
10085711
|
Macherey-Nagel™
718584 |
Cutter Cone Fits |
£34.55
Pack of 5 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
10072981
|
Macherey-Nagel™
718637 |
Connector |
£95.55
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
Description
Stainless steel columns are most frequently used in HPLC. The material is corrosion resistant, pressure stable and easy to work mechanically.