missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute QPCR Mix, SYBR Green, fluorescein

Optimized for SYBR Green chemistry and contains all the components necessary to perform quantitative PCR, with the exception of template and primers.

Brand:  Thermo Scientific™ AB1220A

Product Code. 11842063

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.

ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.

  • Thermo Scientific ABsolute qPCR SYBR Green Mixes are optimized for SYBR Green chemistry and contain all the components necessary to perform quantitative PCR, with the exception of template and primers
  • The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets
  • ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, low ROX concentration, fluorescein or no reference dye
  • A master mix optimized for capillary instruments is also available

Highlights

  • High Sensitivity & Specificity – The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
  • Optimised buffer containing SYBR Green I
  • Reproducibility – All master mixes undergo extensive QC to ensure batch to batch consistency
  • Store at -20°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 1 x 5 mL ABsolute QPCR SYBR Green Fluorescein Mix (2X)
•100 μL MgCl2 (1M)
Sufficient for 400 x 25 μL reactions.

Store at -20°C
High
Taq DNA Polymerase
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
Real Time PCR SYBR Master Mix
Gene Expression
2X
DNA, dsDNA
SYBR
qPCR
Standard
ABsolute
400 Reactions of 25 μL Each
qPCR Cyclers
400 Reactions
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.