Learn More
9-Hydroxyxanthene, 98%, Thermo Scientific Chemicals
£29.50 - £94.50
Chemical Identifiers
CAS | 90-46-0 |
---|---|
Molecular Formula | C13H10O2 |
Molecular Weight (g/mol) | 198.22 |
MDL Number | MFCD00005057 |
InChI Key | JFRMYMMIJXLMBB-UHFFFAOYSA-N |
Synonym | 9-hydroxyxanthene, xanthydrol, xanthanol, 9-xanthydrol, xanthen-9-ol, xanthene, hydroxy, 9-xanthenol, ccris 1640, unii-7131m69ikf, xanthrol |
PubChem CID | 72861 |
IUPAC Name | 9H-xanthen-9-ol |
SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10043350
|
Thermo Scientific Acros
271520050 |
5 g | Glass Bottle |
£29.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10541311
|
Thermo Scientific Acros
271520250 |
25 g | Plastic bottle |
£94.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
90-46-0 | |
198.22 | |
JFRMYMMIJXLMBB-UHFFFAOYSA-N | |
72861 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)O |
C13H10O2 | |
MFCD00005057 | |
9-hydroxyxanthene, xanthydrol, xanthanol, 9-xanthydrol, xanthen-9-ol, xanthene, hydroxy, 9-xanthenol, ccris 1640, unii-7131m69ikf, xanthrol | |
9H-xanthen-9-ol |
Specifications
90-46-0 | |
100.0 | |
White | |
97.5% min. (HPLC) | |
MFCD00005057 | |
9-hydroxyxanthene, xanthydrol, xanthanol, 9-xanthydrol, xanthen-9-ol, xanthene, hydroxy, 9-xanthenol, ccris 1640, unii-7131m69ikf, xanthrol | |
JFRMYMMIJXLMBB-UHFFFAOYSA-N | |
9H-xanthen-9-ol | |
72861 | |
98% | |
9-Hydroxyxanthene, 99% |
97.5 | |
124°C to 127°C | |
Authentic | |
C13H10O2 | |
17, 129 | |
Solubility in water: insoluble | |
C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)O | |
198.22 | |
198.22 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-996-1
missing translation for 'rtecsNumber' : ZD5710000
missing translation for 'tsca' : TSCA
RUO – Research Use Only