missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 8-Channel Filter Plates

For use with Agilent 850-DS dissolution sampling station

Brand:  Cytiva 7707-3600

Code : NEW

Additional Details : Weight : 1.71000kg

 View more versions of this product

Product Code. 15538753

  • £545.00 / Pack of 50
Estimated Shipment: 02-09-2024
to see stock.
Add to basket

Description

Description

  • 0.45μm membrane pore size
  • Available in PTFE, Nylon, PES, and GMF filter media
  • Disposable

Automated dissolution testing

Specifications

Specifications

Agilent 850-DS Dissolution Sampling Station
PES
0.2 μm
Automated dissolution testing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers