missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-4-(trifluoromethyl)coumarin, 98%, Thermo Scientific Chemicals

Catalog Number p-7023211
Click to view available options
Quantity:
1 g
5 g
575-03-1
C10H5F3O3
230.142
MFCD00037578
CCKWMCUOHJAVOL-UHFFFAOYSA-N
7-hydroxy-4-trifluoromethyl coumarin, 7-hydroxy-4-trifluoromethylcoumarin, 7-hydroxy-4-trifluoromethyl-2h-chromen-2-one, 7-hydroxy-4-trifluoromethyl chromen-2-one, 4-trifluoromethyl umbelliferone, 7-hydroxy-4-trifluoromethyl-chromen-2-one, 2h-1-benzopyran-2-one, 7-hydroxy-4-trifluoromethyl, 7,4-hfc, maybridge1_006885, tfmu
5375667
7-hydroxy-4-(trifluoromethyl)chromen-2-one
C1=CC2=C(C=C1O)OC(=O)C=C2C(F)(F)F
575-03-1
C10H5F3O3
230.142
MFCD00037578
CCKWMCUOHJAVOL-UHFFFAOYSA-N
7-hydroxy-4-trifluoromethyl coumarin, 7-hydroxy-4-trifluoromethylcoumarin, 7-hydroxy-4-trifluoromethyl-2h-chromen-2-one, 7-hydroxy-4-trifluoromethyl chromen-2-one, 4-trifluoromethyl umbelliferone, 7-hydroxy-4-trifluoromethyl-chromen-2-one, 2h-1-benzopyran-2-one, 7-hydroxy-4-trifluoromethyl, 7,4-hfc, maybridge1_006885, tfmu
5375667
7-hydroxy-4-(trifluoromethyl)chromen-2-one
C1=CC2=C(C=C1O)OC(=O)C=C2C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 180°C
Quantity 1 g
Formula Weight 230.14
Percent Purity 98%
Chemical Name or Material 7-Hydroxy-4-(trifluoromethyl)coumarin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'