missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 7-Hexadecyn-1-ol, 95%

Product Code. 11439627
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy
Applications
7-Hexadecyn-1-ol is used in the preparation of 15-hexadecynoic acid. It acts as a reagent for pharmaceutical research and in chemical synthesis.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-21-9
Molecular Formula C16H30O
Molecular Weight (g/mol) 238.415
MDL Number MFCD00041709
InChI Key UFLJPANUONDNPT-UHFFFAOYSA-N
Synonym 7-hexadecyn-1-ol, 7-hexadecyne-1-ol, 7-hexadecyn-1-ol,
PubChem CID 549047
IUPAC Name hexadec-7-yn-1-ol
SMILES CCCCCCCCC#CCCCCCCO

Specifications

Linear Formula CH3(CH2)7C≡C(CH2)6OH
Quantity 1 g
Formula Weight 238.41
Percent Purity 95%
Chemical Name or Material 7-Hexadecyn-1-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'