missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoisatin, 97%, Thermo Scientific Chemicals

Product Code. 11384806
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

7-bromoisatin was treated with excess PhMgBr to afford tertiary alcohol 9 in 96 % yield. Isatin oxime triflates undergo a facile fragmentation which is promoted by DBU, to form anthranilonitriles after hydrolytic work-up.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-bromoisatin was treated with excess PhMgBr to afford tertiary alcohol 9 in 96 % yield. Isatin oxime triflates undergo a facile fragmentation which is promoted by DBU, to form anthranilonitriles after hydrolytic work-up.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20780-74-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00774354
InChI Key OCVKSIWBTJCXPV-UHFFFAOYSA-N
Synonym 7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin
PubChem CID 2302353
IUPAC Name 7-bromo-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)Br)NC(=O)C2=O

Specifications

Melting Point 195°C to 199°C
Quantity 10 g
Solubility Information Slightly soluble.
Formula Weight 226.03
Percent Purity 97%
Chemical Name or Material 7-Bromoisatin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'