missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde, Thermo Scientific™
£58.75 - £401.00
Chemical Identifiers
CAS | 898289-54-8 |
---|---|
Molecular Formula | C11H13NO3 |
Molecular Weight (g/mol) | 207.229 |
InChI Key | SBTGZJPHUYLRRU-UHFFFAOYSA-N |
Synonym | 6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yl oxy pyridine-2-carbaldehyde, 6-tetrahydro-2h-pyran-4-yl oxy picolinaldehyde, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde |
PubChem CID | 45594294 |
IUPAC Name | 6-(oxan-4-yloxy)pyridine-2-carbaldehyde |
SMILES | C1COCCC1OC2=CC=CC(=N2)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10481514
|
Thermo Scientific Maybridge
CC58204CB |
250 mg | Amber Glass Bottle |
£58.75
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10491514
|
Thermo Scientific Maybridge
CC58204DA |
1 g | Amber Glass Bottle |
£197.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306244
|
Thermo Scientific Maybridge
CC58204DE |
5 g | Amber Glass Bottle |
£401.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
898289-54-8 | |
207.229 | |
6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yl oxy pyridine-2-carbaldehyde, 6-tetrahydro-2h-pyran-4-yl oxy picolinaldehyde, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde | |
6-(oxan-4-yloxy)pyridine-2-carbaldehyde |
C11H13NO3 | |
SBTGZJPHUYLRRU-UHFFFAOYSA-N | |
45594294 | |
C1COCCC1OC2=CC=CC(=N2)C=O |
Specifications
898289-54-8 | |
6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yl oxy pyridine-2-carbaldehyde, 6-tetrahydro-2h-pyran-4-yl oxy picolinaldehyde, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde | |
C1COCCC1OC2=CC=CC(=N2)C=O | |
207.229 | |
207.23 | |
6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde |
C11H13NO3 | |
SBTGZJPHUYLRRU-UHFFFAOYSA-N | |
6-(oxan-4-yloxy)pyridine-2-carbaldehyde | |
45594294 | |
95% |
Safety and Handling
GHS Signal Word: Warning