Learn More
6-Aminoquinoline, 98%, Thermo Scientific Chemicals
£28.90 - £287.00
Chemical Identifiers
CAS | 580-15-4 |
---|---|
Molecular Formula | C9H8N2 |
Molecular Weight (g/mol) | 144.18 |
InChI Key | RJSRSRITMWVIQT-UHFFFAOYSA-N |
Synonym | 6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine |
PubChem CID | 11373 |
IUPAC Name | quinolin-6-amine |
SMILES | C1=CC2=C(C=CC(=C2)N)N=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15117422
|
Thermo Scientific Acros
|
1 g | Glass bottle |
£28.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15107422
|
Thermo Scientific Acros
369640050 |
5 g | Glass bottle |
£93.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15137422
|
Thermo Scientific Acros
369640250 |
25 g | Glass bottle |
£287.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
580-15-4 | |
144.18 | |
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine | |
quinolin-6-amine |
C9H8N2 | |
RJSRSRITMWVIQT-UHFFFAOYSA-N | |
11373 | |
C1=CC2=C(C=CC(=C2)N)N=C1 |
Specifications
580-15-4 | |
192.0°C to 195.0°C (14.0 mmHg) | |
C9H8N2 | |
6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine | |
RJSRSRITMWVIQT-UHFFFAOYSA-N | |
quinolin-6-amine | |
11373 | |
98% |
115.0°C to 119.0°C | |
97.5% min. (HPLC) | |
22, V,10, 303 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, benzene, diethyl ether,heptane | |
C1=CC2=C(C=CC(=C2)N)N=C1 | |
144.18 | |
144.18 | |
6-Aminoquinoline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-453-
- 6-Aminoquinoline
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only