Learn More
5-(Trifluoromethoxy)isatin, 98%, Thermo Scientific Chemicals
£21.80 - £80.40
Chemical Identifiers
CAS | 169037-23-4 |
---|---|
Molecular Formula | C9H4F3NO3 |
Molecular Weight (g/mol) | 231.13 |
MDL Number | MFCD00192524 |
InChI Key | XHAJMVPMNOBILF-UHFFFAOYSA-N |
Synonym | 5-trifluoromethoxy isatin, 5-trifluoromethoxy indoline-2,3-dione, 5-trifluoromethoxy-1h-indole-2,3-dione, 5-trifluoromethoxyisatin, 1h-indole-2,3-dione, 5-trifluoromethoxy, 5-trifluoromethoxy-2,3-indolinedione, buttpark 34\07-90, acmc-209dz9, d0zr2t |
PubChem CID | 2732752 |
IUPAC Name | 5-(trifluoromethoxy)-1H-indole-2,3-dione |
SMILES | C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=O)N2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11457176
|
Thermo Scientific Alfa Aesar
B25494.03 |
1 g |
£21.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467176
|
Thermo Scientific Alfa Aesar
B25494.06 |
5 g |
£80.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-(Trifluoromethoxy)isatin is used as a reactant for preparation of oxindole derivatives as TAk1 kinase inhibitors and of isatin thiosemicarbazones via condensation with thiosemicarbazones as herpes simplex virus (HSV) inhibitors. It is also used as a reactant for synthesis of 3-substituted 2-indolinone RET kinase inhibitors and for preparation of indolone acetamides as antiseizure agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-(Trifluoromethoxy)isatin is used as a reactant for preparation of oxindole derivatives as TAk1 kinase inhibitors and of isatin thiosemicarbazones via condensation with thiosemicarbazones as herpes simplex virus (HSV) inhibitors. It is also used as a reactant for synthesis of 3-substituted 2-indolinone RET kinase inhibitors and for preparation of indolone acetamides as antiseizure agents.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
169037-23-4 | |
231.13 | |
XHAJMVPMNOBILF-UHFFFAOYSA-N | |
2732752 | |
C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=O)N2 |
C9H4F3NO3 | |
MFCD00192524 | |
5-trifluoromethoxy isatin, 5-trifluoromethoxy indoline-2,3-dione, 5-trifluoromethoxy-1h-indole-2,3-dione, 5-trifluoromethoxyisatin, 1h-indole-2,3-dione, 5-trifluoromethoxy, 5-trifluoromethoxy-2,3-indolinedione, buttpark 34\07-90, acmc-209dz9, d0zr2t | |
5-(trifluoromethoxy)-1H-indole-2,3-dione |
Specifications
169037-23-4 | |
C9H4F3NO3 | |
5-trifluoromethoxy isatin, 5-trifluoromethoxy indoline-2,3-dione, 5-trifluoromethoxy-1h-indole-2,3-dione, 5-trifluoromethoxyisatin, 1h-indole-2,3-dione, 5-trifluoromethoxy, 5-trifluoromethoxy-2,3-indolinedione, buttpark 34\07-90, acmc-209dz9, d0zr2t | |
XHAJMVPMNOBILF-UHFFFAOYSA-N | |
5-(trifluoromethoxy)-1H-indole-2,3-dione | |
2732752 | |
98% |
170°C to 172°C | |
MFCD00192524 | |
Insoluble in water. | |
C1=CC2=C(C=C1OC(F)(F)F)C(=O)C(=O)N2 | |
231.13 | |
231.13 | |
5-(Trifluoromethoxy)isatin |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only