missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitrobenzimidazole, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.
Solubility
Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.
Notes
Incompatible with strong oxidizing agents, strong bases. Store at room temperature.
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.
Solubility
Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.
Notes
Incompatible with strong oxidizing agents, strong bases. Store at room temperature.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 207°C to 211°C |
Quantity | 50 g |
Beilstein | 7926 |
Solubility Information | Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol. |
Formula Weight | 163.14 |
Percent Purity | ≥98% |
Chemical Name or Material | 5-Nitrobenzimidazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'