missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-1H-indole-3-carbaldehyde, 97%, Thermo Scientific™
£26.55 - £443.00
Chemical Identifiers
CAS | 6625-96-3 |
---|---|
Molecular Formula | C9H6N2O3 |
Molecular Weight (g/mol) | 190.158 |
MDL Number | MFCD01313781 |
InChI Key | PHKYMSLVWLYDKP-UHFFFAOYSA-N |
Synonym | 5-nitroindole-3-carbaldehyde, 5-nitroindole-3-carboxaldehyde, 5-nitro-1h-indole-3-carboxaldehyde, pubchem9126, 5-nitro-3-formylindol, 5-nitro-3-formyl-indole, 5-nitro-3-indolealdehyde, 5-nitroindole-3-aldehyde, 5-nitro-3-indolecarbaldehyde, 5-nitro-1h-indole-3-carb aldehyde |
PubChem CID | 246039 |
IUPAC Name | 5-nitro-1H-indole-3-carbaldehyde |
SMILES | C1=CC2=C(C=C1[N+](=O)[O-])C(=CN2)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10161082
|
Thermo Scientific Maybridge
RH01174DA |
1 g |
£26.55
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10121182
|
Thermo Scientific Maybridge
RH01174EA |
10 g |
£155.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10756715
|
Thermo Scientific Maybridge
RH01174EE |
50 g |
£443.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
6625-96-3 | |
190.158 | |
PHKYMSLVWLYDKP-UHFFFAOYSA-N | |
246039 | |
C1=CC2=C(C=C1[N+](=O)[O-])C(=CN2)C=O |
C9H6N2O3 | |
MFCD01313781 | |
5-nitroindole-3-carbaldehyde, 5-nitroindole-3-carboxaldehyde, 5-nitro-1h-indole-3-carboxaldehyde, pubchem9126, 5-nitro-3-formylindol, 5-nitro-3-formyl-indole, 5-nitro-3-indolealdehyde, 5-nitroindole-3-aldehyde, 5-nitro-3-indolecarbaldehyde, 5-nitro-1h-indole-3-carb aldehyde | |
5-nitro-1H-indole-3-carbaldehyde |
Specifications
6625-96-3 | |
97% | |
C9H6N2O3 | |
5-nitroindole-3-carbaldehyde, 5-nitroindole-3-carboxaldehyde, 5-nitro-1h-indole-3-carboxaldehyde, pubchem9126, 5-nitro-3-formylindol, 5-nitro-3-formyl-indole, 5-nitro-3-indolealdehyde, 5-nitroindole-3-aldehyde, 5-nitro-3-indolecarbaldehyde, 5-nitro-1h-indole-3-carb aldehyde | |
C1=CC2=C(C=C1[N+](=O)[O-])C(=CN2)C=O | |
190.158 | |
190.16 | |
5-Nitro-1H-indole-3-carbaldehyde |
97.0 | |
Amber Glass Bottle | |
MFCD01313781 | |
PHKYMSLVWLYDKP-UHFFFAOYSA-N | |
5-nitro-1H-indole-3-carbaldehyde | |
246039 | |
97% |