missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (+)-5-Iodo-2'-deoxyuridine, 98%

Catalog Number p-7081530
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
54-42-2
C9H11IN2O5
354.10
MFCD00134656
XQFRJNBWHJMXHO-RRKCRQDMSA-N
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
5905
CHEBI:147675
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
This item is not returnable. View return policy
54-42-2
C9H11IN2O5
354.10
MFCD00134656
XQFRJNBWHJMXHO-RRKCRQDMSA-N
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
5905
CHEBI:147675
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼190°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 30397
Sensitivity Light sensitive
Merck Index 144891
Formula Weight 354.1
Percent Purity 98%
Chemical Name or Material (+)-5-Iodo-2'-deoxyuridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'