missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroisatin, 98%, Thermo Scientific Chemicals

Product Code. 11435766
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-69-6
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.123
MDL Number MFCD00022795
InChI Key GKODDAXOSGGARJ-UHFFFAOYSA-N
Synonym 5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione
PubChem CID 236566
IUPAC Name 5-fluoro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1F)C(=O)C(=O)N2

Specifications

Melting Point 224°C to 227°C
Quantity 5 g
Beilstein 131241
Formula Weight 165.12
Percent Purity 98%
Chemical Name or Material 5-Fluoroisatin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'