missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals (+)-5-Fluoro-2'-deoxyuridine, 99+%
132.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 148.0°C to 153.0°C |
Color | White |
Loss on Drying | 0.5% (60°C, 4 hrs) (vacuum) max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 100 mg |
Merck Index | 15,4144 |
Specific Rotation Condition | + 35.90 (20.00°C c=1,water) |
Specific Rotation | + 35.90 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- (+)-5-Fluoro-2'-deoxyuridine
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'