missing translation for 'onlineSavingsMsg'
Learn More

5α-Cholestane, 98+%, Thermo Scientific Chemicals

Catalog Number p-3736028
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
481-21-0
C27H48
372.67
MFCD00066412
XIIAYQZJNBULGD-XWLABEFZSA-N
5alpha-cholestane, 5-alpha-cholestane, alpha-cholestane, cholestane, 28,29,30-trinorlanostane, unii-u260hwn305, 5alpha-cholestane 8ci, 5?-cholestane, cholestane van
2723895
CHEBI:35515
(5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCCC4)C)C
This item is not returnable. View return policy
481-21-0
C27H48
372.67
MFCD00066412
XIIAYQZJNBULGD-XWLABEFZSA-N
5alpha-cholestane, 5-alpha-cholestane, alpha-cholestane, cholestane, 28,29,30-trinorlanostane, unii-u260hwn305, 5alpha-cholestane 8ci, 5?-cholestane, cholestane van
2723895
CHEBI:35515
(5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCCC4)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 78°C to 81°C
Color White
Boiling Point 250°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2206
Specific Rotation + 26.5
Solubility Information Solubility in water: insoluble in water. Other solubilities: freely soluble in chloroform (50mg/mL),ether and,slightly soluble in absolute alcohol
Formula Weight 372.67
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 5α-Cholestane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'