missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methylimidazole, 98%, Thermo Scientific™

Product Code. 11372697
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-49-1
Molecular Formula C4H5ClN2
Molecular Weight (g/mol) 116.548
MDL Number MFCD00014505
InChI Key NYDGOZPYEABERA-UHFFFAOYSA-N
Synonym 5-chloro-1-methyl-1h-imidazole, 1-methyl-5-chloroimidazole, 1h-imidazole, 5-chloro-1-methyl, 5-chloro-1-methylimidazole, zlchem 661, pubchem7612, acmc-209qj7, 5-chloro-1-methyl imidazole, 5-chloro-1-methyl-imidazole, ksc494e7d
PubChem CID 70105
IUPAC Name 5-chloro-1-methylimidazole
SMILES CN1C=NC=C1Cl

Specifications

Density 1.25
Boiling Point 89°C to 90°C (14 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.511
Quantity 1 g
Beilstein 110509
Formula Weight 116.55
Percent Purity 98%
Chemical Name or Material 5-Chloro-1-methylimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'