missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol

Product Code. 11440040
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35090-64-3
Molecular Formula C8H9BrO3
Molecular Weight (g/mol) 233.06
MDL Number MFCD02081986
InChI Key UBLQIISPEAEAIX-UHFFFAOYSA-N
Synonym 4-bromo-2-hydroxymethyl-6-methoxyphenol, 4-bromo-2-hydroxymethyl-6-methoxybenzenol, acmc-20amo4, benzenemethanol,5-bromo-2-hydroxy-3-methoxy
PubChem CID 894092
IUPAC Name 4-bromo-2-(hydroxymethyl)-6-methoxyphenol
SMILES COC1=C(O)C(CO)=CC(Br)=C1

Specifications

Quantity 1 g
Formula Weight 233.06
Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'