missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1,3,4-thiadiazol-2-ylamine, 97%, Thermo Scientific™
£18.40 - £130.00
Chemical Identifiers
CAS | 37566-39-5 |
---|---|
Molecular Formula | C2H2BrN3S |
Molecular Weight (g/mol) | 180.02 |
MDL Number | MFCD00464325 |
InChI Key | GLYQQFBHCFPEEU-UHFFFAOYSA-N |
Synonym | 2-amino-5-bromo-1,3,4-thiadiazole, 5-bromo-1,3,4-thiadiazol-2-ylamine, 2-amino-5-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazol-2-amine, 5-bromo, 5-bromo-1,3,4-thiadiazole-2-amine, 5-bromo-1,3,4-thiadiazole-2-ylamine, 5-bromo-1,3,4 thiadiazol-2-ylamine, bromothiadiazolamine, pubchem20972, ksc496o1t |
PubChem CID | 3650883 |
IUPAC Name | 5-bromo-1,3,4-thiadiazol-2-amine |
SMILES | NC1=NN=C(Br)S1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10225504
|
Thermo Scientific Maybridge
MO08007DA |
1 g |
£18.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10663425
|
Thermo Scientific Maybridge
MO08007DE |
5 g |
£74.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10193354
|
Thermo Scientific Maybridge
MO08007EA |
10 g |
£130.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
37566-39-5 | |
180.02 | |
GLYQQFBHCFPEEU-UHFFFAOYSA-N | |
3650883 | |
NC1=NN=C(Br)S1 |
C2H2BrN3S | |
MFCD00464325 | |
2-amino-5-bromo-1,3,4-thiadiazole, 5-bromo-1,3,4-thiadiazol-2-ylamine, 2-amino-5-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazol-2-amine, 5-bromo, 5-bromo-1,3,4-thiadiazole-2-amine, 5-bromo-1,3,4-thiadiazole-2-ylamine, 5-bromo-1,3,4 thiadiazol-2-ylamine, bromothiadiazolamine, pubchem20972, ksc496o1t | |
5-bromo-1,3,4-thiadiazol-2-amine |
Specifications
37566-39-5 | |
C2H2BrN3S | |
2-amino-5-bromo-1,3,4-thiadiazole, 5-bromo-1,3,4-thiadiazol-2-ylamine, 2-amino-5-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazol-2-amine, 5-bromo, 5-bromo-1,3,4-thiadiazole-2-amine, 5-bromo-1,3,4-thiadiazole-2-ylamine, 5-bromo-1,3,4 thiadiazol-2-ylamine, bromothiadiazolamine, pubchem20972, ksc496o1t | |
NC1=NN=C(Br)S1 | |
180.02 | |
180.03 | |
5-Bromo-1, 3, 4-thiadiazol-2-ylamine |
97% | |
MFCD00464325 | |
GLYQQFBHCFPEEU-UHFFFAOYSA-N | |
5-bromo-1,3,4-thiadiazol-2-amine | |
3650883 | |
97% |
Safety and Handling
GHS P Statement Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. Harmful in contact with skin.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Warning