missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-methyl-1H-pyrazole-4-carboxamide, Thermo Scientific™

Product Code. 10600283
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 18213-75-7
Molecular Formula C5H8N4O
Molecular Weight (g/mol) 140.146
MDL Number MFCD00111808
InChI Key JGSQVTVXGXOSCH-UHFFFAOYSA-N
PubChem CID 265696
IUPAC Name 5-amino-1-methylpyrazole-4-carboxamide
SMILES CN1C(=C(C=N1)C(=O)N)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-amino-1-methyl-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide, 5-amino-1-methyl, pubchem22392, 5-amino-1-methyl-4-pyrazolecarboxamide, 5-azanyl-1-methyl-pyrazole-4-carboxamide, 1-methyl-5-amino-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide,5-amino-1-methyl, 5-imino-1-methyl-2h-pyrazole-4-carboxamide, 3-imino-2-methyl-2,3-dihydro-1h-pyrazole-4-carboxamide
Formula Weight 140.15
Percent Purity 95%
Chemical Name or Material 5-Amino-1-methyl-1H-pyrazole-4-carboxamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'