missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Chlorophenyl)-2-furoyl chloride, Thermo Scientific™

Product Code. 11345409
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92973-26-7
Molecular Formula C11H6Cl2O2
Molecular Weight (g/mol) 241.07
MDL Number MFCD02258034
InChI Key DBONYOXUBWINEQ-UHFFFAOYSA-N
Synonym 5-3-chlorophenyl furan-2-carbonyl chloride, 5-3-chlorophenyl-2-furoyl chloride, 5-3-chloro-phenyl-furan-2-carbonyl chloride, acmc-20am9e, 2-furancarbonylchloride, 5-3-chlorophenyl
PubChem CID 3854521
IUPAC Name 5-(3-chlorophenyl)furan-2-carbonyl chloride
SMILES ClC(=O)C1=CC=C(O1)C1=CC(Cl)=CC=C1

Specifications

Quantity 1 g
UN Number UN3261
Formula Weight 241.08
Chemical Name or Material 5-(3-Chlorophenyl)-2-furoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'