missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 406 FX Washer Dispenser

The 406 FX is a compact instrument offering fast, full plate washing along with up to six reagent dispensers. For Research Use Only.

Brand:  Agilent BioTek 406FX-SN

Additional Details : Weight : 20.41166kg

Product Code. 17834677

  • £48435.00 / Each
Estimated Shipment: 29-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Dual-Action manifold optimizes washing for a variety of cell types, in addition to biomagnetic protocols in plate configurations ranging from 96- to 1536-well
  • Non-contact plate dispensing capabilities offer maximum flexibility, including up to four syringe pumps and two peristaltic pump dispensers that enable rapid dispensing of reagents, cells and other precious reagents in volumes ranging from 500 nL to 3000 μL, with minimal dead volume
  • Plate access on both the left and right side of the instrument allows the instrument to be completely automation friendly
  • Flexibility and capabilities of the 406 FX allow non-contact dispensing and full plate washing in a variety of plate densities, all in one instrument
  • Eliminating the need to move microplates between a plate washer and multiple dispensers for complex liquid handling workflows
  • Multiple independent fluid paths means that up to six different reagents can be loaded and dispensed in a single procedure, eliminating the need to incorporate multiple dispensers
  • Touchscreen interface makes protocol creation simple and intuitive
  • Left-side and right-side robotic arm accessibility allows the 406 FX to be easily integrated with the Agilent BioStack™ and other third-party automation, to increase assay throughput
  • Direct-to-drain waste system eliminates the need to empty waste bottles as waste can go directly to a drain
  • Combination of angled dispense tubes, highly adjustable aspiration and dispense rates, and x- and y-axis positioning ensure the integrity of cell monolayers during dispensing and wash procedures
  • Built-in Ultrasonic Advantage allows the instrument to clean buildup in the manifold tube automatically, preventing failure during wash procedures
  • Magnet accessories automate biomagnetic separation procedures
  • Computer control, protocol creation, and automation interfacing is available with the optional LHC software
  • LHC Secure software can computer control the 406 FX and provides features for compliance to 21CFR Part 11

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Specifications

Specifications

±1 to 5%
500 nL to 3000 μL/well (Peristaltic Pump), 3 to 3000 μL/well (Syringe Pump)
≤2 μL/well
Programmable in minutes and seconds, up to 60 min.
20.4 kg
Color Touchscreen
Yes
25 in.
12.5 in.
14.25 in.
45 lb.
Dispenser Bottles (4 L, 10 L, 20 L)
96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates
13 to 36 sec.
Microplate Washer/Dispenser
1 to 250
50/60 Hz
96/384 Dual-action Wash Manifold, Ultrasonic Advantage, Internal 4-wash Buffer Switching, Primary Peristaltic Pump
63.5 cm
31.8 cm
36.2 cm
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.