Learn More
Masterflex™ 4-way Stopcock
Use these stopcocks for a variety of applications. Cole-Parmer™ 4-way Stopcocks feature a PC body and an HDPE stem.
Brand: Masterflex™ 30600-03
Code : A0
Additional Details : Weight : 0.02880kg
Description
Use these stopcocks for a variety of applications. Cole-Parmer™ 4-way Stopcocks feature a PC body and an HDPE stem.
- Four-way stopcock
- Choose stopcocks with either a male luer lock or a male luer slip
- Handle rotates 360 degrees on four-way stopcocks
Specifications
Stopcock | |
Polycarbonate and HDPE | |
Male Luer Slip |
Polycarbonate, HDPE | |
Male Luer Lock or A Male Luer Slip |