missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals

Product Code. 15425398
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68755-42-0
Molecular Formula C10H7F3O
Molecular Weight (g/mol) 200.16
MDL Number MFCD07772121
InChI Key LJVBFMQEZSEGRL-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1-indanone, 4-trifluoromethyl-2,3-dihydro-1h-inden-1-one, 4-trifluoromethyl-2,3-dihydroinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-trifluoromethyl, 4-trifluoromethyl indan-1-one, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one, 2,3-dihydro-1-oxo-4-trifluoromethyl-1h-indene, acmc-1b8x3, 2,3-dihydro-4-trifluoromethyl-1h-inden-1-one
PubChem CID 21921093
IUPAC Name 4-(trifluoromethyl)-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C(=CC=C2)C(F)(F)F

Specifications

Melting Point 38°C to 42°C
Boiling Point 67°C to 72°C (0.1 mmHg)
Flash Point >108°C (226°F)
Quantity 5 g
Formula Weight 200.16
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)-1-indanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'