Learn More
4-Pyrimidin-2-ylbenzoyl chloride, TECH, Thermo Scientific™
£38.00 - £89.70
Chemical Identifiers
CAS | 679806-84-9 |
---|---|
Molecular Formula | C11H7ClN2O |
Molecular Weight (g/mol) | 218.64 |
MDL Number | MFCD09064974 |
InChI Key | AQFRTZZTRGGBKO-UHFFFAOYSA-N |
Synonym | 4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride |
PubChem CID | 21092945 |
IUPAC Name | 4-pyrimidin-2-ylbenzoyl chloride |
SMILES | C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10679233
|
Thermo Scientific Maybridge
CC56202CB |
250 mg | Amber Glass Bottle |
£38.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10613065
|
Thermo Scientific Maybridge
CC56202DA |
1 g | Amber Glass Bottle |
£89.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
679806-84-9 | |
218.64 | |
AQFRTZZTRGGBKO-UHFFFAOYSA-N | |
21092945 | |
C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl |
C11H7ClN2O | |
MFCD09064974 | |
4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride | |
4-pyrimidin-2-ylbenzoyl chloride |
Specifications
679806-84-9 | |
100.0 | |
C11H7ClN2O | |
4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride | |
C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl | |
218.64 | |
218.64 | |
4-Pyrimidin-2-ylbenzoyl chloride |
89.5 | |
90% | |
MFCD09064974 | |
AQFRTZZTRGGBKO-UHFFFAOYSA-N | |
4-pyrimidin-2-ylbenzoyl chloride | |
21092945 | |
90% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger