missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcarbamoylbenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15479917
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330793-45-8
Molecular Formula C13H12BNO3
Molecular Weight (g/mol) 241.053
MDL Number MFCD04115683
InChI Key DAUSGSRIYCUJDK-UHFFFAOYSA-N
Synonym 4-phenylcarbamoyl phenyl boronic acid, 4-phenylcarbamoyl phenylboronic acid, 4-phenylaminocarbonylphenylboronic acid, 4-phenylaminocarbonylphenyl boronic acid, 4-phenylaminocarbonyl benzeneboronic acid, 4-phenylamino-1-carbonyl phenyl boronic acid, n-phenyl 4-boronobenzamide, acmc-1afql, 4-phenylcarbamoylbenzeneboronic acid
PubChem CID 11096747
IUPAC Name [4-(phenylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2)(O)O

Specifications

Melting Point 210°C to 216°C
Quantity 1 g
Formula Weight 241.05
Percent Purity 97%
Chemical Name or Material 4-Phenylcarbamoylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'