missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Octyloxybenzaldehyde, 97%, Thermo Scientific Chemicals
£37.30 - £392.00
Chemical Identifiers
CAS | 24083-13-4 |
---|---|
Molecular Formula | C15H22O2 |
Molecular Weight (g/mol) | 234.339 |
MDL Number | MFCD00014136 |
InChI Key | KVOWZHASDIKNFK-UHFFFAOYSA-N |
Synonym | 4-octyloxy benzaldehyde, 4-octyloxybenzaldehyde, 4-n-octyloxybenzaldehyde, p-octyloxybenzaldehyde, benzaldehyde, 4-octyloxy, p-octyloxy benzaldehyde, p-n-octoxy benzaldehyde, p-n-octyloxybenzaldehyde, acmc-209g8t |
PubChem CID | 90358 |
IUPAC Name | 4-octoxybenzaldehyde |
SMILES | CCCCCCCCOC1=CC=C(C=C1)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405536
|
Thermo Scientific Alfa Aesar
B20692.06 |
5 g |
£37.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415536
|
Thermo Scientific Alfa Aesar
B20692.14 |
25 g |
£139.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425536
|
Thermo Scientific Alfa Aesar
B20692.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24083-13-4 | |
234.339 | |
KVOWZHASDIKNFK-UHFFFAOYSA-N | |
90358 | |
CCCCCCCCOC1=CC=C(C=C1)C=O |
C15H22O2 | |
MFCD00014136 | |
4-octyloxy benzaldehyde, 4-octyloxybenzaldehyde, 4-n-octyloxybenzaldehyde, p-octyloxybenzaldehyde, benzaldehyde, 4-octyloxy, p-octyloxy benzaldehyde, p-n-octoxy benzaldehyde, p-n-octyloxybenzaldehyde, acmc-209g8t | |
4-octoxybenzaldehyde |
Specifications
24083-13-4 | |
140°C to 142°C (0.1 mmHg) | |
1.522 | |
1913284 | |
4-octyloxy benzaldehyde, 4-octyloxybenzaldehyde, 4-n-octyloxybenzaldehyde, p-octyloxybenzaldehyde, benzaldehyde, 4-octyloxy, p-octyloxy benzaldehyde, p-n-octoxy benzaldehyde, p-n-octyloxybenzaldehyde, acmc-209g8t | |
CCCCCCCCOC1=CC=C(C=C1)C=O | |
234.339 | |
234.33 | |
4-n-Octyloxybenzaldehyde |
0.982 | |
C15H22O2 | |
MFCD00014136 | |
Air Sensitive | |
KVOWZHASDIKNFK-UHFFFAOYSA-N | |
4-octoxybenzaldehyde | |
90358 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 246-012-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only