Learn More
4'-Hydroxy-2'-methylacetophenone, 97%, Thermo Scientific Chemicals
£40.90 - £421.00
Chemical Identifiers
CAS | 875-59-2 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.18 |
MDL Number | MFCD00002303 |
InChI Key | IAMNVCJECQWBLZ-UHFFFAOYSA-N |
PubChem CID | 70133 |
ChEBI | CHEBI:87314 |
IUPAC Name | 1-(4-hydroxy-2-methylphenyl)ethanone |
SMILES | CC(=O)C1=CC=C(O)C=C1C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10147903
|
Thermo Scientific Acros
121390050 |
5 g | Glass bottle |
£40.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10588470
|
Thermo Scientific Acros
121390250 |
25 g | Glass bottle |
£137.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10331624
|
Thermo Scientific Acros
121391000 |
100 g | Glass bottle |
£421.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
875-59-2 | |
150.18 | |
IAMNVCJECQWBLZ-UHFFFAOYSA-N | |
CHEBI:87314 | |
CC(=O)C1=CC=C(O)C=C1C |
C9H10O2 | |
MFCD00002303 | |
70133 | |
1-(4-hydroxy-2-methylphenyl)ethanone |
Specifications
875-59-2 | |
100.0 | |
Tan to White | |
Authentic | |
C9H10O2 | |
MFCD00002303 | |
4'-hydroxy-2'-methylacetophenone, 1-4-hydroxy-2-methylphenyl ethanone, 4-hydroxy-2-methylacetophenone, 2-methyl-4-hydroxyacetophenone, ethanone, 1-4-hydroxy-2-methylphenyl, 3-methyl-4-acetylphenol, 1-4-hydroxy-2-methylphenyl ethan-1-one, 1-acetyl-4-hydroxy-2-methylbenzene, ethanone,1-4-hydroxy-2-methylphenyl, 2'-methyl-4'-hydroxyacetophenone | |
IAMNVCJECQWBLZ-UHFFFAOYSA-N | |
1-(4-hydroxy-2-methylphenyl)ethanone | |
70133 | |
150.18 | |
Crystals or Powder |
96.0 | |
127°C to 132°C | |
313°C | |
96% min. (HPLC) | |
HOC6H3(CH3)COCH3 | |
08, 111 | |
Solubility in water: .. Other solubilities: soluble in methanol | |
CC(=O)C1=CC=C(O)C=C1C | |
150.18 | |
CHEBI:87314 | |
97% | |
4'-Hydroxy-2'-methylacetophenone |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-874-2
RUO – Research Use Only