missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 15421258
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-59-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00017942
InChI Key VLWRKVBQUANIGI-UHFFFAOYSA-N
PubChem CID 68024
IUPAC Name 4-fluoro-N-methylaniline
SMILES CNC1=CC=C(C=C1)F

Specifications

Density 1.04
Boiling Point 79°C (11 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.532
Quantity 5 g
UN Number UN2810
Synonym benzenamine, 4-fluoro-n-methyl, unii-88f75yis0f, n-methyl-4-fluoroaniline, acmc-1aem5, 4-fluoro-n-methyl aniline, 4-fluoro-n-methyl-aniline, 4-fluoro-n-methylbenzenamine, 4-fluorophenyl-methylamine, benzenamine,4-fluoro-n-methyl
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'